Learn More
1,5-Cyclooctadiene, 99%, stabilized
CAS: 111-78-4 | C8H12 | 108.18 g/mol
Supplier: Thermo Scientific Chemicals 297120010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1, 5-Cyclooctadiene | |
| 111-78-4 | |
| 100.0 | |
| Colorless | |
| 151°C (101.3 kPa) | |
| Authentic | |
| Glass bottle | |
| 1.4920 to 1.4940 | |
| 1 L | |
| 0.88 | |
| 1,5-cyclooctadiene, cycloocta-1,5-diene, 1z,5z-cycloocta-1,5-diene, 1,5-cyclooctadiene z,z, 1z,5z-cyclooctadiene, z,z-cycloocta-1,5-diene, 1-cis,5-cis-cyclooctadiene, unii-1e1vvd385z, cis,cis-cycloocta-1,5-diene, 1,5-cod | |
| VYXHVRARDIDEHS-QGTKBVGQSA-N | |
| (5Z)-cycloocta-1,5-diene | |
| 50 to 150ppm t-butylcatechol | |
| 1.3 mPa.s (20°C) | |
| 99% |
| Stabilized | |
| 98.5 | |
| -69.5°C | |
| 0.8800g/mL | |
| 38°C | |
| 98.5% min. (GC) | |
| C8H12 | |
| MFCD00001752 | |
| 05, 116 | |
| 15, 2730 | |
| Solubility in water: 780mg/l (20°C). Other solubilities: soluble in benzene and carbon tetrachloride | |
| C1C\C=C/CC\C=C/1 | |
| 108.18 | |
| 10937607 | |
| 108.18 | |
| Liquid |
Chemical Identifiers
| 111-78-4 | |
| 108.18 | |
| VYXHVRARDIDEHS-QGTKBVGQSA-N | |
| 10937607 |
| C8H12 | |
| MFCD00001752 | |
| 1,5-cyclooctadiene, cycloocta-1,5-diene, 1z,5z-cycloocta-1,5-diene, 1,5-cyclooctadiene z,z, 1z,5z-cyclooctadiene, z,z-cycloocta-1,5-diene, 1-cis,5-cis-cyclooctadiene, unii-1e1vvd385z, cis,cis-cycloocta-1,5-diene, 1,5-cod | |
| C1C\C=C/CC\C=C/1 |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a
GHS Signal Word: Danger
EINECSNumber : 203-907-1
RUO – Research Use Only