missing translation for 'onlineSavingsMsg'
Learn More

1,5-Anhydro-D-sorbitol, 97%

Catalog No. p-7077824
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459912500 250 mg
AC459910010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459912500 Supplier Thermo Scientific Chemicals Supplier No. 459912500
Only null left

CAS: 154-58-5 | C6H12O5 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 154-58-5
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
InChI Key MPCAJMNYNOGXPB-SLPGGIOYSA-N
Synonym 1,5-anhydro-d-glucitol, 1,5-anhydroglucitol, 1,5-anhydrosorbitol, 1,5-anhydro-d-sorbitol, 1-deoxy-d-glucopyranose, aceritol, 1-deoxy-d-glucose, d-glucitol, 1,5-anhydro, unii-54bb3b7xmz, glucitol, 1,5-anhydro
PubChem CID 64960
ChEBI CHEBI:16070
IUPAC Name (2R,3S,4R,5S)-2-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1C(C(C(C(O1)CO)O)O)O

Specifications

Melting Point 141.0°C to 144.0°C
Infrared Spectrum Authentic
Quantity 250 mg
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material 1,5-Anhydro-D-sorbitol
EINECSNumber 205-829-3

RUO – Research Use Only