missing translation for 'onlineSavingsMsg'
Learn More

1,5,9-Decatriene 95.0+%, TCI America™

Catalog No. p-7127054
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19935ML 5 mL
D199325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D19935ML Supplier TCI America Supplier No. D19935ML
Only null left

Chemical Identifiers

CAS 13393-64-1
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00055625
InChI Key GMGYODAMPOELNZ-MDZDMXLPSA-N
Synonym 1,5,9-decatriene, 5e-deca-1,5,9-triene, 1,5,9-decatriene, mixture of cis and trans, e-1,5,9-decatriene, 5e-1,5,9-decatriene, 5e-1,5,9-decatriene #, 1,5,9-decatriene, cis + trans
PubChem CID 5362636
IUPAC Name (5E)-deca-1,5,9-triene
SMILES C=CCC\C=C\CCC=C

Specifications

Color Yellow
Boiling Point 55°C
Quantity 5 mL
UN Number 3295
Formula Weight 136.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,5,9-Decatriene