CAS RN 13393-64-1
IUPAC Name:
(5E)-deca-1,5,9-triene
Synonyms:
(5E)-deca-1,5,9-triene
Molecular Weight (g/mol):
136.24
Molecular Formula:
C10H16
InChi Key:
GMGYODAMPOELNZ-MDZDMXLPSA-N
SMILES:
C=CCC\C=C\CCC=C
1
–
1
of
1
results
1
1,5,9-Decatriene 95.0+%, TCI America™
CAS: 13393-64-1 Molecular Formula: C10H16 Molecular Weight (g/mol): 136.24 MDL Number: MFCD00055625 InChI Key: GMGYODAMPOELNZ-MDZDMXLPSA-N Synonym: 1,5,9-decatriene,5e-deca-1,5,9-triene,1,5,9-decatriene, mixture of cis and trans,e-1,5,9-decatriene,5e-1,5,9-decatriene,5e-1,5,9-decatriene #,1,5,9-decatriene, cis + trans PubChem CID: 5362636 IUPAC Name: (5E)-deca-1,5,9-triene SMILES: C=CCC\C=C\CCC=C