missing translation for 'onlineSavingsMsg'
Learn More

1,5,6,7-Tetrahydro-2-methyl-4H-indol-4-one 98.0+%, TCI America™

Catalog No. p-7137185
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23765G 5 g
T237625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23765G Supplier TCI America Supplier No. T23765G

Chemical Identifiers

CAS 35308-68-0
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD04972500
InChI Key HJIYEKHYUGHTAC-UHFFFAOYSA-N
Synonym 2-Methyl-4-oxo-4,5,6,7-tetrahydroindole
PubChem CID 9815223
IUPAC Name 2-methyl-4,5,6,7-tetrahydro-1H-indol-4-one
SMILES CC1=CC2=C(CCCC2=O)N1

Specifications

Melting Point 204°C
Quantity 5 g
Formula Weight 149.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,5,6,7-Tetrahydro-2-methyl-4H-indol-4-one