accessibility menu, dialog, popup

UserName

CAS RN 35308-68-0

IUPAC Name: 2-methyl-4,5,6,7-tetrahydro-1H-indol-4-one
Synonyms: 2-methyl-1,5,6,7-tetrahydroindol-4-one
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: HJIYEKHYUGHTAC-UHFFFAOYSA-N
SMILES: CC1=CC2=C(CCCC2=O)N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results