missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Trifluoromethylphenyl)piperazine 98.0+%, TCI America™
Supplier: TCI America T274125G
Specifications
| 1-(4-Trifluoromethylphenyl)piperazine | |
| 90°C | |
| C11H13F3N2 | |
| 25 g | |
| IBQMAPSJLHRQPE-UHFFFAOYSA-N | |
| 1-[4-(trifluoromethyl)phenyl]piperazine | |
| 121718 | |
| ≥98.0% (GC,T) |
| 30459-17-7 | |
| White-Yellow | |
| MFCD00040765 | |
| 1-4-trifluoromethylphenyl piperazine, 1-4-trifluoromethyl phenyl piperazine, piperazine, 1-4-trifluoromethyl phenyl, 1-4-trifluoromethylphenyl-piperazine, 1-4-trifluoromethyl-phenyl-piperazine, n-4-trifluoromethylphenyl piperazine, 4-trifluoromethyl phenyl piperazine, zlchem 639, acmc-209hfy | |
| C1CN(CCN1)C2=CC=C(C=C2)C(F)(F)F | |
| 230.234 | |
| 230.23 | |
| Crystalline Powder |
Chemical Identifiers
| 30459-17-7 | |
| 230.234 | |
| IBQMAPSJLHRQPE-UHFFFAOYSA-N | |
| 121718 | |
| C1CN(CCN1)C2=CC=C(C=C2)C(F)(F)F |
| C11H13F3N2 | |
| MFCD00040765 | |
| 1-4-trifluoromethylphenyl piperazine, 1-4-trifluoromethyl phenyl piperazine, piperazine, 1-4-trifluoromethyl phenyl, 1-4-trifluoromethylphenyl-piperazine, 1-4-trifluoromethyl-phenyl-piperazine, n-4-trifluoromethylphenyl piperazine, 4-trifluoromethyl phenyl piperazine, zlchem 639, acmc-209hfy | |
| 1-[4-(trifluoromethyl)phenyl]piperazine |
Safety and Handling
TSCA : No