missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Trifluoromethylphenyl)piperazine 98.0+%, TCI America™
$134.75 - $301.48
Chemical Identifiers
| CAS | 30459-17-7 |
|---|---|
| Molecular Formula | C11H13F3N2 |
| Molecular Weight (g/mol) | 230.234 |
| MDL Number | MFCD00040765 |
| InChI Key | IBQMAPSJLHRQPE-UHFFFAOYSA-N |
| Synonym | 1-4-trifluoromethylphenyl piperazine, 1-4-trifluoromethyl phenyl piperazine, piperazine, 1-4-trifluoromethyl phenyl, 1-4-trifluoromethylphenyl-piperazine, 1-4-trifluoromethyl-phenyl-piperazine, n-4-trifluoromethylphenyl piperazine, 4-trifluoromethyl phenyl piperazine, zlchem 639, acmc-209hfy |
| PubChem CID | 121718 |
| IUPAC Name | 1-[4-(trifluoromethyl)phenyl]piperazine |
| SMILES | C1CN(CCN1)C2=CC=C(C=C2)C(F)(F)F |
Chemical Identifiers
| 30459-17-7 | |
| 230.234 | |
| IBQMAPSJLHRQPE-UHFFFAOYSA-N | |
| 121718 | |
| C1CN(CCN1)C2=CC=C(C=C2)C(F)(F)F |
| C11H13F3N2 | |
| MFCD00040765 | |
| 1-4-trifluoromethylphenyl piperazine, 1-4-trifluoromethyl phenyl piperazine, piperazine, 1-4-trifluoromethyl phenyl, 1-4-trifluoromethylphenyl-piperazine, 1-4-trifluoromethyl-phenyl-piperazine, n-4-trifluoromethylphenyl piperazine, 4-trifluoromethyl phenyl piperazine, zlchem 639, acmc-209hfy | |
| 1-[4-(trifluoromethyl)phenyl]piperazine |
Specifications
| 30459-17-7 | |
| White-Yellow | |
| MFCD00040765 | |
| 1-4-trifluoromethylphenyl piperazine, 1-4-trifluoromethyl phenyl piperazine, piperazine, 1-4-trifluoromethyl phenyl, 1-4-trifluoromethylphenyl-piperazine, 1-4-trifluoromethyl-phenyl-piperazine, n-4-trifluoromethylphenyl piperazine, 4-trifluoromethyl phenyl piperazine, zlchem 639, acmc-209hfy | |
| C1CN(CCN1)C2=CC=C(C=C2)C(F)(F)F | |
| 230.234 | |
| 230.23 | |
| Crystalline Powder |
| 90°C | |
| C11H13F3N2 | |
| 5 g | |
| IBQMAPSJLHRQPE-UHFFFAOYSA-N | |
| 1-[4-(trifluoromethyl)phenyl]piperazine | |
| 121718 | |
| ≥98.0% (GC,T) | |
| 1-(4-Trifluoromethylphenyl)piperazine |
Safety and Handling
TSCA : No