missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Trifluoromethylbenzyl)piperazine 97.0+%, TCI America™

Catalog No. T30751G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30751G 1 g
T30755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30751G Supplier TCI America Supplier No. T30751G
Only null left

Chemical Identifiers

CAS 107890-32-4
Molecular Formula C12H17F3N2
Molecular Weight (g/mol) 246.28
MDL Number MFCD03407490
InChI Key FAFAFWFQFVLXGF-UHFFFAOYSA-P
PubChem CID 2782803
IUPAC Name 4-{[4-(trifluoromethyl)phenyl]methyl}piperazine-1,4-diium
SMILES FC(F)(F)C1=CC=C(C[NH+]2CC[NH2+]CC2)C=C1

Specifications

Color Yellow
Boiling Point 100°C
Quantity 1 g
Formula Weight 244.26
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-(4-Trifluoromethylbenzyl)piperazine