missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(Trifluoromethyl)phenyl]ethanol, 96%

Catalog No. p-7046943
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1687803 1 g
AAL1687806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1687803 Supplier Thermo Scientific Chemicals Supplier No. L1687803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1737-26-4 | C9H9F3O | 190.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1737-26-4
Molecular Formula C9H9F3O
Molecular Weight (g/mol) 190.165
MDL Number MFCD00039618
InChI Key YMXIDIAEXNLCFT-UHFFFAOYSA-N
Synonym 1-4-trifluoromethyl phenyl ethanol, 1-4-trifluoromethyl phenyl ethan-1-ol, a-methyl-4-trifluoromethylbenzyl alcohol, alpha-methyl-4-trifluoromethylbenzyl alcohol, alpha-methyl-4-trifluoromethyl benzyl alcohol, acmc-1boxb, 1-4-trifluoromethylphenyl-ethanol, 1-4-trifluoromethyl-phenyl-ethanol, alpha-methyl-4 trifluoromethyl benzyl alcohol
PubChem CID 137186
IUPAC Name 1-[4-(trifluoromethyl)phenyl]ethanol
SMILES CC(C1=CC=C(C=C1)C(F)(F)F)O

Specifications

Density 1.237
Boiling Point 62°C to 63°C (0.5 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.458
Quantity 1 g
Beilstein 2257808
Formula Weight 190.17
Percent Purity 97%
Chemical Name or Material 1-[4-(Trifluoromethyl)phenyl]ethanol
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only