missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| 1,4-Thioxane | |
| Yellow | |
| C4H8OS | |
| 25 g | |
| 1,4-thioxane, p-thioxane, 1-oxa-4-thiacyclohexane, ccris 1746, 1,4-oxathiin, 2,3,5,6-tetrahydro, thioather, 1 pound not4-thioxane, 1-thia-4-oxacyclohexane, acmc-1bt7b, wln: t6o dstj | |
| C1CSCCO1 | |
| 104.167 | |
| 104.17 | |
| Liquid |
| 15980-15-1 | |
| 67°C | |
| MFCD00006573 | |
| 1993 | |
| JBYHSSAVUBIJMK-UHFFFAOYSA-N | |
| 1,4-oxathiane | |
| 27596 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 15980-15-1 | |
| 104.167 | |
| JBYHSSAVUBIJMK-UHFFFAOYSA-N | |
| 27596 | |
| C1CSCCO1 |
| C4H8OS | |
| MFCD00006573 | |
| 1,4-thioxane, p-thioxane, 1-oxa-4-thiacyclohexane, ccris 1746, 1,4-oxathiin, 2,3,5,6-tetrahydro, thioather, 1 pound not4-thioxane, 1-thia-4-oxacyclohexane, acmc-1bt7b, wln: t6o dstj | |
| 1,4-oxathiane |
Safety and Handling
RTECSNumber : RP4200000
TSCA : Yes