missing translation for 'onlineSavingsMsg'
Learn More

1,4-Thioxane 97.0+%, TCI America™

Catalog No. p-7136828
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15435G 5 g
T154325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15435G Supplier TCI America Supplier No. T15435G
Only null left

Chemical Identifiers

CAS 15980-15-1
Molecular Formula C4H8OS
Molecular Weight (g/mol) 104.167
MDL Number MFCD00006573
InChI Key JBYHSSAVUBIJMK-UHFFFAOYSA-N
Synonym 1,4-thioxane, p-thioxane, 1-oxa-4-thiacyclohexane, ccris 1746, 1,4-oxathiin, 2,3,5,6-tetrahydro, thioather, 1 pound not4-thioxane, 1-thia-4-oxacyclohexane, acmc-1bt7b, wln: t6o dstj
PubChem CID 27596
IUPAC Name 1,4-oxathiane
SMILES C1CSCCO1

Specifications

Color Yellow
Boiling Point 67°C
Quantity 5 g
UN Number 1993
Formula Weight 104.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Thioxane
RTECSNumber RP4200000
TSCA Yes