missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Pyridyl)pyridinium Chloride Hydrochloride Hydrate 98.0+%, TCI America™

Catalog No. p-7134679
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P05655G 5 g
P056525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P05655G Supplier TCI America Supplier No. P05655G

Chemical Identifiers

CAS 5421-92-1
Molecular Formula C10H10Cl2N2
Molecular Weight (g/mol) 229.104
MDL Number MFCD00012821
InChI Key QZOFFMRDIRXGKJ-UHFFFAOYSA-M
Synonym 1-4-pyridyl pyridinium chloride hydrochloride, 1,4'-bipyridin-1-ium chloride hydrochloride, n-4-pyridyl pyridinium chloride hydrochloride, 1-4-pyridinyl pyridinium chloride hydrochloride, 1,4'-bipyridinium, chloride, 1,4'-bipyridinium, chloride, hydrochloride 1:1:1, 1,4'-bipyridinium, chloride, hydrochloride, 1-4-pyridyl pyridinium chloride hcl, 4-pyridin-1-ium-1-ylpyridine chloride hydrochloride, 4-pyridylpyridine, chloride, chloride
PubChem CID 79461
IUPAC Name 4-pyridin-1-ium-1-ylpyridine;chloride;hydrochloride
SMILES C1=CC=[N+](C=C1)C2=CC=NC=C2.Cl.[Cl-]

Specifications

Color Yellow
Quantity 5 g
Formula Weight 229.10
Percent Purity ≥98.0% (HPLC,T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Pyridyl)pyridinium Chloride Hydrochloride Hydrate