accessibility menu, dialog, popup

UserName

CAS RN 5421-92-1

IUPAC Name: hydrogen 1λ⁵-[1,4'-bipyridin]-1-ylium dichloride
Synonyms: hydrogen 1λ⁵-[1,4'-bipyridin]-1-ylium dichloride
Molecular Weight (g/mol): 229.1
Molecular Formula: C10H10Cl2N2
InChi Key: QZOFFMRDIRXGKJ-UHFFFAOYSA-M
SMILES: [H+].[Cl-].[Cl-].C1=CC=[N+](C=C1)C1=CC=NC=C1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results