missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Pyridyl)pyridinium Chloride Hydrochloride Hydrate 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 g |
| Formula Weight | 229.10 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Grade | Laboratory |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-(4-Pyridyl)pyridinium Chloride Hydrochloride Hydrate |
Safety and Handling
| TSCA | Yes |