missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Phenylenediamine 98.0+%, TCI America™
Supplier: TCI America P0170250G
Specifications
| 1,4-Phenylenediamine | |
| 142°C | |
| 267°C | |
| MFCD00007901 | |
| 1673 | |
| CBCKQZAAMUWICA-UHFFFAOYSA-N | |
| benzene-1,4-diamine | |
| 7814 | |
| 108.14 | |
| Crystalline Powder |
| 106-50-3 | |
| Red-White | |
| C6H8N2 | |
| 250 g | |
| p-phenylenediamine, 1,4-benzenediamine, 1,4-diaminobenzene, 1,4-phenylenediamine, 4-aminoaniline, para-phenylenediamine, p-aminoaniline, p-diaminobenzene, orsin, 4-phenylenediamine | |
| C1=CC(=CC=C1N)N | |
| 108.144 | |
| CHEBI:51403 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 106-50-3 | |
| 108.144 | |
| CBCKQZAAMUWICA-UHFFFAOYSA-N | |
| 7814 | |
| benzene-1,4-diamine |
| C6H8N2 | |
| MFCD00007901 | |
| p-phenylenediamine, 1,4-benzenediamine, 1,4-diaminobenzene, 1,4-phenylenediamine, 4-aminoaniline, para-phenylenediamine, p-aminoaniline, p-diaminobenzene, orsin, 4-phenylenediamine | |
| CHEBI:51403 | |
| C1=CC(=CC=C1N)N |
Safety and Handling
EINECSNumber : (3)-0185&(5)-4998
RTECSNumber : SS8050000
TSCA : Yes