missing translation for 'onlineSavingsMsg'
Learn More

1,4-Phenylenediamine 98.0+%, TCI America™

Catalog No. p-7134466
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P017025G 25 g
P0170250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P017025G Supplier TCI America Supplier No. P017025G
Only null left

Chemical Identifiers

CAS 106-50-3
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00007901
InChI Key CBCKQZAAMUWICA-UHFFFAOYSA-N
Synonym p-phenylenediamine, 1,4-benzenediamine, 1,4-diaminobenzene, 1,4-phenylenediamine, 4-aminoaniline, para-phenylenediamine, p-aminoaniline, p-diaminobenzene, orsin, 4-phenylenediamine
PubChem CID 7814
ChEBI CHEBI:51403
IUPAC Name benzene-1,4-diamine
SMILES C1=CC(=CC=C1N)N

Specifications

Melting Point 142°C
Color Red-White
Boiling Point 267°C
Quantity 25 g
UN Number 1673
Formula Weight 108.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Phenylenediamine
EINECSNumber (3)-0185&(5)-4998
RTECSNumber SS8050000
TSCA Yes