missing translation for 'onlineSavingsMsg'
Learn More

1,4-Phenylenediacetonitrile, 97%

Catalog No. AAL1346414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1346414 25 g
AAL1346406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1346414 Supplier Thermo Scientific Chemicals Supplier No. L1346414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 622-75-3 | C10H8N2 | 156.188 g/mol

1,4-Phenylenediacetonitrile, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Phenylenediacetonitrile, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 622-75-3
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.188
MDL Number MFCD00001923
InChI Key FUQCKESKNZBNOG-UHFFFAOYSA-N
Synonym 1,4-phenylenediacetonitrile, 1,4-benzenediacetonitrile, p-xylylene dicyanide, p-phenylenediacetonitrile, p-benzenediacetonitrile, 2,2'-1,4-phenylene diacetonitrile, p-bis cyanomethyl benzene, 1,4-bis cyanomethyl benzene, p-cyanomethyl benzyl cyanide, 2-4-cyanomethyl phenyl acetonitrile
PubChem CID 69327
IUPAC Name 2-[4-(cyanomethyl)phenyl]acetonitrile
SMILES C1=CC(=CC=C1CC#N)CC#N

Specifications

Melting Point 94°C to 98°C
Quantity 25 g
UN Number UN3439
Beilstein 1866157
Solubility Information Insoluble in water.
Formula Weight 156.19
Percent Purity 97%
Chemical Name or Material 1,4-Phenylenediacetonitrile
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement GHS P Statement
P280h-P309-P310
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 210-751-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only