missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Phenylene Diisothiocyanate 98.0+%, TCI America™
Supplier: TCI America P127925G
Specifications
| 1,4-Phenylene Diisothiocyanate | |
| 130°C | |
| C8H4N2S2 | |
| 25 g | |
| bitoscanate, 1,4-phenylene diisothiocyanate, jonit, p-phenylene diisothiocyanate, benzene, 1,4-diisothiocyanato, bitoscanat, bitoscanate inn, phenylene thiocyanate, bitoscanatum latin, 1,4-phenylenediisothiocyanate | |
| C1=CC(=CC=C1N=C=S)N=C=S | |
| 192.254 | |
| 192.25 | |
| Crystalline Powder |
| 4044-65-9 | |
| White-Yellow | |
| MFCD00004811 | |
| 2811 | |
| OMWQUXGVXQELIX-UHFFFAOYSA-N | |
| 1,4-diisothiocyanatobenzene | |
| 19958 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 4044-65-9 | |
| 192.254 | |
| OMWQUXGVXQELIX-UHFFFAOYSA-N | |
| 19958 | |
| C1=CC(=CC=C1N=C=S)N=C=S |
| C8H4N2S2 | |
| MFCD00004811 | |
| bitoscanate, 1,4-phenylene diisothiocyanate, jonit, p-phenylene diisothiocyanate, benzene, 1,4-diisothiocyanato, bitoscanat, bitoscanate inn, phenylene thiocyanate, bitoscanatum latin, 1,4-phenylenediisothiocyanate | |
| 1,4-diisothiocyanatobenzene |
Safety and Handling
RTECSNumber : NX9150000
TSCA : Yes