missing translation for 'onlineSavingsMsg'
Learn More

1,4-Phenylene Diisothiocyanate 98.0+%, TCI America™

Catalog No. P12795G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12795G 5 g
P127925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12795G Supplier TCI America Supplier No. P12795G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4044-65-9
Molecular Formula C8H4N2S2
Molecular Weight (g/mol) 192.254
MDL Number MFCD00004811
InChI Key OMWQUXGVXQELIX-UHFFFAOYSA-N
Synonym bitoscanate, 1,4-phenylene diisothiocyanate, jonit, p-phenylene diisothiocyanate, benzene, 1,4-diisothiocyanato, bitoscanat, bitoscanate inn, phenylene thiocyanate, bitoscanatum latin, 1,4-phenylenediisothiocyanate
PubChem CID 19958
IUPAC Name 1,4-diisothiocyanatobenzene
SMILES C1=CC(=CC=C1N=C=S)N=C=S

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 192.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Phenylene Diisothiocyanate
RTECSNumber NX9150000
TSCA Yes