missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Nitrophenyl)glycerol, 99%

Catalog No. AAL1996803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1996803 1 g
AAL1996806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1996803 Supplier Thermo Scientific Chemicals Supplier No. L1996803

CAS: 2207-68-3 | C9H11NO5 | 213.189 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2207-68-3
Molecular Formula C9H11NO5
Molecular Weight (g/mol) 213.189
MDL Number MFCD00017037
InChI Key IUZVZBIQZKBWCC-UHFFFAOYSA-N
Synonym 1-4-nitrophenyl propane-1,2,3-triol, p-nitrophenylglycerol, 1-4-nitrophenyl glycerol, 1-4-nitrophenyl-1,2,3-propanetriol, 1,2,3-propanetriol, 1-4-nitrophenyl, 1-4-nitrophenyl glycerol pnpg, 4-nitrophenylglycerol, pubchem9886, para-nitrophenylglycerol, maybridge1_002064
PubChem CID 102764
IUPAC Name 1-(4-nitrophenyl)propane-1,2,3-triol
SMILES C1=CC(=CC=C1C(C(CO)O)O)[N+](=O)[O-]

Specifications

Melting Point 95°C to 97°C
Odor Odorless
Quantity 1 g
Beilstein 7376091
Sensitivity Hygroscopic
Formula Weight 213.19
Percent Purity 99%
Chemical Name or Material 1-(4-Nitrophenyl)glycerol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-624-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only