missing translation for 'onlineSavingsMsg'
Learn More

1,4-Naphthoquinone 98.0+%, TCI America™

Catalog No. N0040500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0040500G 500 g
N004025G 25 g
N0040100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N0040500G Supplier TCI America Supplier No. N0040500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 130-15-4
Molecular Formula C10H6O2
Molecular Weight (g/mol) 158.156
MDL Number MFCD00001676
InChI Key FRASJONUBLZVQX-UHFFFAOYSA-N
Synonym 1,4-naphthoquinone, 1,4-naphthalenedione, p-naphthoquinone, naphthoquinone, alpha-naphthoquinone, 1,4-naphthylquinone, usaf cy-10, 1,4-dihydronaphthalene-1,4-dione, 1,4-dihydro-1,4-diketonaphthalene, 1,4-naftochinon
PubChem CID 8530
ChEBI CHEBI:27418
IUPAC Name naphthalene-1,4-dione
SMILES C1=CC=C2C(=O)C=CC(=O)C2=C1

Specifications

Melting Point 125°C
Quantity 500 g
UN Number 2811
Formula Weight 158.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Naphthoquinone
EINECSNumber (4)-0372
RTECSNumber QL7175000
TSCA Yes