missing translation for 'onlineSavingsMsg'
Learn More

1,4-Naphthoquinone, 97+% (dry wt.), cont. up to 5% water

CAS: 130-15-4 | C10H6O2 | 158.156 g/mol

Supplier:  Thermo Scientific Chemicals A109580E

 View more versions of this product

Catalog No. AAA109580E


Only null left
Explore available promotions

Description

Description

1,4-Naphthoquinone is used as a strong dienophile in Diels-Alder reaction. It is also used as a precursor to anthroquinone and 5-nitro-1,4-naphthalenedione, which finds application in the preparation of aminoanthroquinone and is also used as a dye precursor. It is a basic component of Vitamin K.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Naphthoquinone is used as a strong dienophile in Diels-Alder reaction. It is also used as a precursor to anthroquinone and 5-nitro-1,4-naphthalenedione, which finds application in the preparation of aminoanthroquinone and is also used as a dye precursor. It is a basic component of Vitamin K.

Solubility
Soluble in alkali hydroxide solvents, sparingly soluble in petroleum ether and water. Freely soluble in carbondioxide, acetic acid, benzene and choloroform.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong reducing agents.
Specifications

Specifications

1,4-Naphthoquinone
120°C to 124°C
142°C (288°F)
(dry wt.)
MFCD00001676
UN2811
14,6395
Soluble in alkali hydroxide solvents,sparingly soluble in petroleum ether and water. Freely soluble in carbondioxide,acetic acid,benzene and choloroform.
C1=CC=C2C(=O)C=CC(=O)C2=C1
158.156
CHEBI:27418
≥97%
130-15-4
1.42
Irritating
C10H6O2
2500 g
878524
1,4-naphthoquinone, 1,4-naphthalenedione, p-naphthoquinone, naphthoquinone, alpha-naphthoquinone, 1,4-naphthylquinone, usaf cy-10, 1,4-dihydronaphthalene-1,4-dione, 1,4-dihydro-1,4-diketonaphthalene, 1,4-naftochinon
FRASJONUBLZVQX-UHFFFAOYSA-N
naphthalene-1,4-dione
8530
158.15
Contains up to 5% water

Chemical Identifiers

130-15-4
158.156
FRASJONUBLZVQX-UHFFFAOYSA-N
8530
naphthalene-1,4-dione
C10H6O2
MFCD00001676
1,4-naphthoquinone, 1,4-naphthalenedione, p-naphthoquinone, naphthoquinone, alpha-naphthoquinone, 1,4-naphthylquinone, usaf cy-10, 1,4-dihydronaphthalene-1,4-dione, 1,4-dihydro-1,4-diketonaphthalene, 1,4-naftochinon
CHEBI:27418
C1=CC=C2C(=O)C=CC(=O)C2=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H330-H315-H319-H317-H335
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P260-P264b-P270-P271-P272-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H301+H311-H314-H317-H330-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 204-977-6

RTECSNumber : QL7175000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only