missing translation for 'onlineSavingsMsg'
Learn More

1,4-Naphthalenedicarboxylic Acid 95.0+%, TCI America™

Catalog No. p-7133891
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N060625G 25 g
N0606100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N060625G Supplier TCI America Supplier No. N060625G
Only null left

Chemical Identifiers

CAS 605-70-9
Molecular Formula C12H8O4
Molecular Weight (g/mol) 216.19
MDL Number MFCD00014312
InChI Key ABMFBCRYHDZLRD-UHFFFAOYSA-N
Synonym 1,4-naphthalenedicarboxylic acid, 1,4-naphthalenedicarboxylicacid, 1,4-naphthalicacid, 1,4-naphthalic acid, pubchem21317, acmc-1b0c7, ksc357a6b, naphthalene-1,4-dicarboxylicacid, naphthalene-1,4-dicarboxlic acid, naphthaline-1,4-dicarboxylic acid
PubChem CID 69065
IUPAC Name naphthalene-1,4-dicarboxylic acid
SMILES OC(=O)C1=C2C=CC=CC2=C(C=C1)C(O)=O

Specifications

Melting Point 320°C
Quantity 25 g
Formula Weight 216.19
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Naphthalenedicarboxylic Acid
EINECSNumber (4)-1765
TSCA Yes