missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methylphenyl)piperazine, 98%, Thermo Scientific™

Chemical Identifiers

CAS 39593-08-3
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.26
MDL Number MFCD00040737
InChI Key ONEYFZXGNFNRJH-UHFFFAOYSA-N
Synonym 1-4-methylphenyl piperazine, 1-p-tolyl piperazine, 1-p-tolyl-piperazine, 1-4-methylphenyl-piperazine, 1-4-methyl phenyl piperazine, 1-p-tolylpiperazine, acmc-1adtx, 4-methylphenyl piperazine, 4-4-methylphenyl piperazine, 1-4-methyl-phenyl-piperazine
PubChem CID 83113
IUPAC Name 1-(4-methylphenyl)piperazine
SMILES CC1=CC=C(C=C1)N2CCNCC2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC370830050
SDS View Documents
Thermo Scientific Chemicals
370830050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

39593-08-3
176.26
ONEYFZXGNFNRJH-UHFFFAOYSA-N
83113
CC1=CC=C(C=C1)N2CCNCC2
C11H16N2
MFCD00040737
1-4-methylphenyl piperazine, 1-p-tolyl piperazine, 1-p-tolyl-piperazine, 1-4-methylphenyl-piperazine, 1-4-methyl phenyl piperazine, 1-p-tolylpiperazine, acmc-1adtx, 4-methylphenyl piperazine, 4-4-methylphenyl piperazine, 1-4-methyl-phenyl-piperazine
1-(4-methylphenyl)piperazine

Specifications

39593-08-3
100.0
97.5% min. (GC)
C11H16N2
5g
ONEYFZXGNFNRJH-UHFFFAOYSA-N
1-(4-methylphenyl)piperazine
83113
98%
97.5
Authentic
Glass bottle
MFCD00040737
1-4-methylphenyl piperazine, 1-p-tolyl piperazine, 1-p-tolyl-piperazine, 1-4-methylphenyl-piperazine, 1-4-methyl phenyl piperazine, 1-p-tolylpiperazine, acmc-1adtx, 4-methylphenyl piperazine, 4-4-methylphenyl piperazine, 1-4-methyl-phenyl-piperazine
CC1=CC=C(C=C1)N2CCNCC2
176.26
176.26
1-(4-Methylphenyl)piperazine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Danger

EINECSNumber : 254-534-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only