missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Iodophenyl)pyrrole, 97%

Catalog No. AAA1530506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1530506 5 g
AAA1530503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1530506 Supplier Thermo Scientific Chemicals Supplier No. A1530506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 92636-36-7 | C10H8IN | 269.085 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 92636-36-7
Molecular Formula C10H8IN
Molecular Weight (g/mol) 269.085
MDL Number MFCD00052399
InChI Key FMURNAZHVQDQQN-UHFFFAOYSA-N
Synonym 1-4-iodophenyl pyrrole, 1-4-iodophenyl-1h-pyrrole, acmc-20apaj, 4-iodophenyl pyrrole, maybridge1_002926, 1-4-iodo-phenyl-1h-pyrrole, 1h-pyrrole,1-4-iodophenyl, 1-4-iodophenyl-1h-pyrrole #
PubChem CID 272430
IUPAC Name 1-(4-iodophenyl)pyrrole
SMILES C1=CN(C=C1)C2=CC=C(C=C2)I

Specifications

Melting Point 128°C to 133°C
Quantity 5 g
Beilstein 7915717
Sensitivity Light sensitive
Formula Weight 269.09
Percent Purity 97%
Chemical Name or Material 1-(4-Iodophenyl)pyrrole
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only