accessibility menu, dialog, popup

UserName

CAS RN 92636-36-7

IUPAC Name: 1-(4-iodophenyl)-1H-pyrrole
Synonyms: 1-(4-iodophenyl)pyrrole
Molecular Weight (g/mol): 269.09
Molecular Formula: C10H8IN
InChi Key: FMURNAZHVQDQQN-UHFFFAOYSA-N
SMILES: IC1=CC=C(C=C1)N1C=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results