Learn More
MilliporeSigma™ Supelco™ 1/4 in. Brass Swagelok™ and PTFE Ferrule End-Fitting Assembly, Precleaned
Catalog No. 111009534
Change view
Click to view available options
For Use With (Equipment):
1/4 in. (6.35 mm) Thermal Desorption Tube
Catalog No. For Use With (Equipment)
111009534 1/4 in. (6.35 mm) Thermal Desorption Tube
1 options
Catalog No. 111009534 Supplier MilliporeSigma™ Supelco™ Supplier No. 23094U
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ideal for those who purchase unconditioned glass and stainless steel thermal desorption tubes and condition in their own lab.
The assembly includes a 1/4 in. brass Swagelok™™ and 1/4 in. PTFE ferrule, pre-cleaned and ready for use.
Specifications
Material Brass (Precleaned and Assembled)
Quantity 20/Pk.
For Use With (Equipment) 1/4 in. (6.35 mm) Thermal Desorption Tube
Swagelok is a registered trademark of Swagelok Company