missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Hydroxyphenyl)-5-mercaptotetrazole, 96%

Catalog No. AAB2200306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2200306 5 g
AAB2200314 25 g
AAB2200322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2200306 Supplier Thermo Scientific Chemicals Supplier No. B2200306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 52431-78-4 | C7H6N4OS | 194.212 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52431-78-4
Molecular Formula C7H6N4OS
Molecular Weight (g/mol) 194.212
MDL Number MFCD00132898
InChI Key MOXZSKYLLSPATM-UHFFFAOYSA-N
Synonym 1-4-hydroxyphenyl-5-mercaptotetrazole, 1-4-hydroxyphenyl-1h-tetrazole-5-thiol, 1-4-hydroxyphenyl-2h-tetrazole-5-thione, 1-4-hydroxyphenyl-5-mercapto-1h-tetrazole, 5h-tetrazole-5-thione, 1,2-dihydro-1-4-hydroxyphenyl, 1-4-hydroxyphenyl-1h-tetrazole-5 4h-thione, 4-5-sulfanyl-1h-1,2,3,4-tetrazol-1-yl phenol, 1,2-dihydro-1-p-hydroxyphenyl-5h-tetrazole-5-thione, 4-5-mercapto-1h-tetrazol-1-yl phenol, 4-5-sulfanyl-1h-tetrazol-1-yl phenol
PubChem CID 3034725
IUPAC Name 1-(4-hydroxyphenyl)-2H-tetrazole-5-thione
SMILES C1=CC(=CC=C1N2C(=S)N=NN2)O

Specifications

Melting Point 169°C to 171°C
Quantity 5 g
Formula Weight 194.21
Percent Purity 96%
Chemical Name or Material 1-(4-Hydroxyphenyl)-5-mercaptotetrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 257-909-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only