missing translation for 'onlineSavingsMsg'
Learn More

1-[(4-Fluorophenyl)carbamoyl]cyclopropanecarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7130059
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10831G 1 g
F10835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10831G Supplier TCI America Supplier No. F10831G
Only null left

Chemical Identifiers

CAS 849217-48-7
Molecular Formula C11H10FNO3
Molecular Weight (g/mol) 223.203
MDL Number MFCD11226313
InChI Key PFMAFXYUHZDKPY-UHFFFAOYSA-N
PubChem CID 21081530
IUPAC Name 1-[(4-fluorophenyl)carbamoyl]cyclopropane-1-carboxylic acid
SMILES C1CC1(C(=O)NC2=CC=C(C=C2)F)C(=O)O

Specifications

Melting Point 175°C
Color White
Quantity 1 g
Formula Weight 223.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-[(4-Fluorophenyl)carbamoyl]cyclopropanecarboxylic Acid