accessibility menu, dialog, popup

UserName

CAS RN 849217-48-7

IUPAC Name: 1-[(4-fluorophenyl)carbamoyl]cyclopropane-1-carboxylic acid
Synonyms: 1-[(4-fluorophenyl)carbamoyl]cyclopropane-1-carboxylic acid
Molecular Weight (g/mol): 223.2
Molecular Formula: C11H10FNO3
InChi Key: PFMAFXYUHZDKPY-UHFFFAOYSA-N
SMILES: OC(=O)C1(CC1)C(=O)NC1=CC=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results