missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride, 97%, Thermo Scientific™

Catalog No. CC00119DA
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC00119DA 1g
CC00119DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC00119DA Supplier Thermo Scientific Chemicals Supplier No. CC00119DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 423768-49-4
Molecular Formula C11H8ClFN2O
Molecular Weight (g/mol) 238.646
MDL Number MFCD03659694
InChI Key INIAJXQDJISNJB-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl-5-methyl-1h-pyrazole-4-carbonyl chloride, 1-4-fluorophenyl-5-methylpyrazole-4-carbonyl chloride, 1-4-fluorophenyl-5-methyl-4-pyrazolecarbonyl chloride, 1-4-fluorophenyl-5-methyl-pyrazole-4-carbonyl chloride
PubChem CID 2779893
IUPAC Name 1-(4-fluorophenyl)-5-methylpyrazole-4-carbonyl chloride
SMILES CC1=C(C=NN1C2=CC=C(C=C2)F)C(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1g
Formula Weight 238.65
Percent Purity 97%
Chemical Name or Material 1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.