missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluorophenyl)-2-methyl-2-propylamine, 96%

Catalog No. AAL1185803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1185803 1 g
AAL1185806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1185803 Supplier Thermo Scientific Chemicals Supplier No. L1185803

CAS: 1200-27-7 | C10H14FN | 167.227 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1200-27-7
Molecular Formula C10H14FN
Molecular Weight (g/mol) 167.227
MDL Number MFCD00082702
InChI Key JITFIYFVPMQJOK-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl-2-methylpropan-2-amine, 1-4-fluorophenyl-2-methyl-2-propylamine, 4-fluorophenyl-2-methyl-2-aminopropane, 1-4-fluorophenyl-2-methyl-2-aminopropane, 1-4-fluorophenyl-2-methyl-2-propanamine, 1-4-fluorophenyl-2-methylprop-2-ylamine, 1-4-fluorophenyl-2-methyl-propan-2-amine, 1,1-dimethyl-2-4-fluorophenyl ethylamine, 2-4-fluoro-phenyl-1,1-dimethyl-ethylamine, acmc-1alz4
PubChem CID 518471
IUPAC Name 1-(4-fluorophenyl)-2-methylpropan-2-amine
SMILES CC(C)(CC1=CC=C(C=C1)F)N

Specifications

Boiling Point 82°C to 83°C (9 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.495
Quantity 1 g
UN Number UN2735
Beilstein 2802368
Formula Weight 167.23
Percent Purity 96%
Chemical Name or Material 1-(4-Fluorophenyl)-2-methyl-2-propylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only