missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluorobenzyl)-1,4-diazepane, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals AW00512DE

Quantity 5g

 View more versions of this product

Catalog No. AW00512DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

76141-89-4
208.28
KZCHYFUMQQWGJK-UHFFFAOYSA-N
2774433
C1CNCCN(C1)CC2=CC=C(C=C2)F
C12H17FN2
MFCD00816588
1-4-fluorobenzyl-1,4-diazepane, 1-4-fluorobenzyl homopiperazine, 1-4-fluorophenyl methyl-1,4-diazepane, 1-4-fluoro-benzyl-1,4 diazepane, 1-4-fluorophenyl methyl-1,4-diazaperhydroepine, acmc-1bluy, 1-4-fluorobenzyl-1,4 diazepane
1-[(4-fluorophenyl)methyl]-1,4-diazepane

Specifications

1-(4-Fluorobenzyl)-1, 4-diazepane
94.5
95%
5g
1-4-fluorobenzyl-1,4-diazepane, 1-4-fluorobenzyl homopiperazine, 1-4-fluorophenyl methyl-1,4-diazepane, 1-4-fluoro-benzyl-1,4 diazepane, 1-4-fluorophenyl methyl-1,4-diazaperhydroepine, acmc-1bluy, 1-4-fluorobenzyl-1,4 diazepane
C1CNCCN(C1)CC2=CC=C(C=C2)F
208.28
208.28
76141-89-4
100.0
C12H17FN2
MFCD00816588
KZCHYFUMQQWGJK-UHFFFAOYSA-N
1-[(4-fluorophenyl)methyl]-1,4-diazepane
2774433
95%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents