missing translation for 'onlineSavingsMsg'
Learn More

1,4-Epoxycyclohexane 98.0+%, TCI America™

Supplier:  TCI America E050825ML

 View more versions of this product

Catalog No. E050825ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1,4-Epoxycyclohexane
Yellow
25 mL
C6H10O
1993
YPWFNLSXQIGJCK-UHFFFAOYSA-N
7-oxabicyclo[2.2.1]heptane
9234
≥98.0% (GC)
279-49-2
120°C
Clear Liquid at 20°C
MFCD00082342
1,4-epoxycyclohexane, 7-oxabicyclo 2.2.1 heptane, 7-oxanorbornane, 1,4-oxycyclohexane, 3,6-endoxocyclohexane, 3,6-endooxycyclohexane, cyclohexane,4-epoxy, cyclohexane,6-endoxo, cyclohexane, 1,4-epoxy
C1CC2CCC1O2
98.15
98.15

Chemical Identifiers

279-49-2
98.15
YPWFNLSXQIGJCK-UHFFFAOYSA-N
9234
C1CC2CCC1O2
C6H10O
MFCD00082342
1,4-epoxycyclohexane, 7-oxabicyclo 2.2.1 heptane, 7-oxanorbornane, 1,4-oxycyclohexane, 3,6-endoxocyclohexane, 3,6-endooxycyclohexane, cyclohexane,4-epoxy, cyclohexane,6-endoxo, cyclohexane, 1,4-epoxy
7-oxabicyclo[2.2.1]heptane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions