missing translation for 'onlineSavingsMsg'
Learn More

1,4-Epoxycyclohexane 98.0+%, TCI America™

Catalog No. p-7129037
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05085ML 5 mL
E050825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05085ML Supplier TCI America Supplier No. E05085ML
Only null left

Chemical Identifiers

CAS 279-49-2
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00082342
InChI Key YPWFNLSXQIGJCK-UHFFFAOYSA-N
Synonym 1,4-epoxycyclohexane, 7-oxabicyclo 2.2.1 heptane, 7-oxanorbornane, 1,4-oxycyclohexane, 3,6-endoxocyclohexane, 3,6-endooxycyclohexane, cyclohexane,4-epoxy, cyclohexane,6-endoxo, cyclohexane, 1,4-epoxy
PubChem CID 9234
IUPAC Name 7-oxabicyclo[2.2.1]heptane
SMILES C1CC2CCC1O2

Specifications

Color Yellow
Quantity 5 mL
Physical Form Clear Liquid at 20°C
UN Number 1993
Boiling Point 120°C
Formula Weight 98.15
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1,4-Epoxycyclohexane