missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dithio-DL-threitol, Electrophoresis Grade, 99%

CAS: 3483-12-3 | C4H10O2S2 | 154.24 g/mol

Supplier:  Thermo Scientific Chemicals J6454506

 View more versions of this product

Catalog No. AAJ6454506


Only null left
Explore available promotions

Description

Description

1,4-Dithio-DL-threitol is used as a protective agent for preventing oxidation of thiol groups and reagent for the reduction of disulfide groups in proteins. It is employed to investigate a-ketoglutarate dehydrogenase function, flavin cofactors in proteins and protein moieties involved in peroxide reduction. It is also useful for stabilizing sulfhydryl-containing enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dithio-DL-threitol is used as a protective agent for preventing oxidation of thiol groups and reagent for the reduction of disulfide groups in proteins. It is employed to investigate a-ketoglutarate dehydrogenase function, flavin cofactors in proteins and protein moieties involved in peroxide reduction. It is also useful for stabilizing sulfhydryl-containing enzymes.

Solubility
Soluble in water, ethanol, chloroform, diethyl ether, acetone, ether and ethyl acetate.

Notes
Air and heat sensitive. Hygroscopic. Incompatible with strong oxidizing agents, reducing agents and alkalies.
Specifications

Specifications

1,4-Dithio-DL-threitol
40°C to 43°C
125°C to 130°C (12 mmHg)
Unpleasant
MFCD00004877
1719757
14,3376
Soluble in water,ethanol,chloroform,diethyl ether,acetone,ether and ethyl acetate.
C(C(C(CS)O)O)S
154.24
CHEBI:42170
99%
Powder
12-3-3483
White
>110°C (230°F)
C4H10O2S2
5 g
Air sensitive
dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
VHJLVAABSRFDPM-IMJSIDKUSA-N
(2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
446094
154.25
Electrophoresis

Chemical Identifiers

12-3-3483
154.24
VHJLVAABSRFDPM-IMJSIDKUSA-N
446094
(2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
C4H10O2S2
MFCD00004877
dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
CHEBI:42170
C(C(C(CS)O)O)S
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 222-468-7

RTECSNumber : EK1610000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only