missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diphenyl-2-butyne-1,4-dione 96.0+%, TCI America™

Catalog No. p-7128412
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
250 mg
Catalog No. Quantity
D4699250MG 250 mg
D46991G 1 g
2 options

Catalog No. D4699250MG

Supplier: TCI America D4699250MG

Only null left

Chemical Identifiers

CAS 1087-09-8
Molecular Formula C16H10O2
Molecular Weight (g/mol) 234.254
InChI Key BLCJVQFFSUNDMV-UHFFFAOYSA-N
Synonym Dibenzoylacetylene
PubChem CID 227980
IUPAC Name 1,4-diphenylbut-2-yne-1,4-dione
SMILES C1=CC=C(C=C1)C(=O)C#CC(=O)C2=CC=CC=C2

Specifications

Melting Point 113°C
Color Yellow
Quantity 250 mg
Formula Weight 234.25
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Diphenyl-2-butyne-1,4-dione
TSCA No
Recommended Storage Refrigerator