missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diphenoxybenzene 98.0+%, TCI America™

Catalog No. D20175G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20175G 5 g
D201725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20175G Supplier TCI America Supplier No. D20175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3061-36-7
Molecular Formula C18H14O2
Molecular Weight (g/mol) 262.308
MDL Number MFCD00038368
InChI Key UVGPELGZPWDPFP-UHFFFAOYSA-N
Synonym benzene, 1,4-diphenoxy, p-diphenoxybenzene, hydroquinone diphenyl ether, p-phenoxyphenoxybenzene, benzene, p-diphenoxy, 4-phenoxydiphenyl oxide, 1,1'-1,4-phenylenebis oxy dibenzene, 1,4-diphenoxy-benzene, acmc-1cqa0
PubChem CID 520487
ChEBI CHEBI:39271
IUPAC Name 1,4-diphenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)OC3=CC=CC=C3

Specifications

Melting Point 76°C
Color White-Yellow
Boiling Point 182°C
Quantity 5 g
Formula Weight 262.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Diphenoxybenzene