missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diphenoxybenzene, 97%

CAS: 3061-36-7 | C18H14O2 | 262.308 g/mol

Supplier:  Thermo Scientific Chemicals A1486414

 View more versions of this product

Catalog No. AAA1486414

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,4-Diphenoxybenzene
72°C to 73°C
172°C (0.01 mmHg)
C18H14O2
25 g
benzene, 1,4-diphenoxy, p-diphenoxybenzene, hydroquinone diphenyl ether, p-phenoxyphenoxybenzene, benzene, p-diphenoxy, 4-phenoxydiphenyl oxide, 1,1'-1,4-phenylenebis oxy dibenzene, 1,4-diphenoxy-benzene, acmc-1cqa0
C1=CC=C(C=C1)OC2=CC=C(C=C2)OC3=CC=CC=C3
262.308
CHEBI:39271
98%
3061-36-7
1.083
202°C (395°F)
MFCD00038368
2215945
UVGPELGZPWDPFP-UHFFFAOYSA-N
1,4-diphenoxybenzene
520487
262.31

Chemical Identifiers

3061-36-7
262.308
UVGPELGZPWDPFP-UHFFFAOYSA-N
520487
1,4-diphenoxybenzene
C18H14O2
MFCD00038368
benzene, 1,4-diphenoxy, p-diphenoxybenzene, hydroquinone diphenyl ether, p-phenoxyphenoxybenzene, benzene, p-diphenoxy, 4-phenoxydiphenyl oxide, 1,1'-1,4-phenylenebis oxy dibenzene, 1,4-diphenoxy-benzene, acmc-1cqa0
CHEBI:39271
C1=CC=C(C=C1)OC2=CC=C(C=C2)OC3=CC=CC=C3
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 000-000-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only