missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diphenoxybenzene, 97%

Catalog No. AAA1486422
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1486422 100 g
AAA1486406 5 g
AAA1486414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1486422 Supplier Thermo Scientific Chemicals Supplier No. A1486422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3061-36-7 | C18H14O2 | 262.308 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3061-36-7
Molecular Formula C18H14O2
Molecular Weight (g/mol) 262.308
MDL Number MFCD00038368
InChI Key UVGPELGZPWDPFP-UHFFFAOYSA-N
Synonym benzene, 1,4-diphenoxy, p-diphenoxybenzene, hydroquinone diphenyl ether, p-phenoxyphenoxybenzene, benzene, p-diphenoxy, 4-phenoxydiphenyl oxide, 1,1'-1,4-phenylenebis oxy dibenzene, 1,4-diphenoxy-benzene, acmc-1cqa0
PubChem CID 520487
ChEBI CHEBI:39271
IUPAC Name 1,4-diphenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)OC3=CC=CC=C3

Specifications

Melting Point 72°C to 73°C
Density 1.083
Boiling Point 172°C (0.01 mmHg)
Flash Point 202°C (395°F)
Quantity 100 g
Beilstein 2215945
Formula Weight 262.31
Percent Purity 98%
Chemical Name or Material 1,4-Diphenoxybenzene
EINECSNumber 000-000-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only