missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane Solution NMR Reference Standard, 40% in benzene-d6 (99.6 atom % D), NMR Tube Size: 3 mm 8 in., MilliporeSigma™ Supelco™

Catalog No. 111018510
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018510 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018510 Supplier Merck Emd Millipore Supplier No. 6121111EA

Specifications

Packaging NMR Tube Size: 3 mm 8 in.
CAS 123-91-1
Concentration 40% in benzene-d 6 (99.6 atom% D)
Linear Formula C4H8O2
MDL Number MFCD00006571
Quantity 1 Ea.
Formula Weight 88.11
Grade NMR Reference Standard