missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane Solution, NMR Reference Standard, 40% in benzene-d6 (99.6 atom% D), MilliporeSigma™ Supelco™

Catalog No. 111018273
Change view
Click to view available options
Quantity:
0.7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018273 0.7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018273 Supplier Merck Emd Millipore Supplier No. 7361718IN

Specifications

Packaging 5 mm O.D. tube
CAS 123-91-1
Flash Point 12°C - Closed Cup
Linear Formula C4H8O2
MDL Number MFCD00006571
Type NMR Reference Standard
Quantity 0.7 mL
Formula Weight 88.11
Grade NMR Reference Standard
Concentration or Composition (by Analyte or Components) 40% in benzene-d 6 (99.6 atom % D)