missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dioxane, CHROMASOLV™ Plus, for HPLC, ≥99.5%, Honeywell Riedel-de Haën™
CHROMASOLV™ Plus, for HPLC,≥99.5%
$1,441.58 - $5,180.25
Chemical Identifiers
| CAS | 123-91-1 |
|---|---|
| Molecular Formula | C4H8O2 |
| MDL Number | MFCD00006571 |
| InChI Key | RYHBNJHYFVUHQT-UHFFFAOYSA-N |
| PubChem CID | 31275 |
| ChEBI | CHEBI:47032 |
| IUPAC Name | 1,4-dioxane |
| SMILES | C1COCCO1 |
Description
- For HPLC, spectrophotometry, and environmental analysis
- Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
- Low UV impurity profile
- Low impurity gradient LC baseline
- Very low levels of non-volatile impurities
Chemical Identifiers
| 123-91-1 | |
| MFCD00006571 | |
| 31275 | |
| 1,4-dioxane |
| C4H8O2 | |
| RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
| CHEBI:47032 | |
| C1COCCO1 |
Specifications
| 10°C to 12°C (literature) | |
| max. 0.01 at 295 nm, max. 0.10 at 270 nm, max. 0.20 at 250 nm, max. 0.50 at 235 nm, max. 0.70 at 220 nm, max. 0.01 at 400 nm, max. 0.01 at 300 nm | |
| 1.034Â g/mLÂ at 25Â °C(lit.) | |
| 2.5 L | |
| Narrow Mouth Glass Bottle | |
| C4H8O2 | |
| UN1165 | |
| dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin | |
| C1COCCO1 | |
| 88.106 | |
| CHEBI:47032 | |
| ≥99.5% | |
| 1,4-Dioxane, CHROMASOLV™ Plus, for HPLC, ≥99.5% |
| 123-91-1 | |
| 100.0000% | |
| 100°C to 102°C | |
| 11°C | |
| C4H8O2 | |
| MFCD00006571 | |
| 102551 | |
| RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
| 1,4-dioxane | |
| 31275 | |
| 88.11 | |
| HPLC |
Safety and Handling
ShelfLife : 720 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Dioxane
EINECSNumber : 204-661-8
Recommended Storage : Room Temp