missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane-2,5-dione, 97%

Catalog No. p-7024857
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AAB2009606 5 g
AAB2009614 25 g
2 options

Catalog No. AAB2009606

Supplier: Thermo Scientific Chemicals B2009606

Only null left

CAS: 502-97-6 | C4H4O4 | 116.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 502-97-6
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00081108
InChI Key RKDVKSZUMVYZHH-UHFFFAOYSA-N
Synonym glycolide, glycollide, unii-yrz676pgu6, p-dioxane-2,5-dione, yrz676pgu6, acmc-1aybf, 1,4-dioxan-2,5-dione, 2,5-dioxo-1,4-dioxane, p-dioxane-2,5-dione 8ci
PubChem CID 65432
IUPAC Name 1,4-dioxane-2,5-dione
SMILES O=C1COC(=O)CO1

Specifications

Melting Point 81°C to 83°C
Quantity 5 g
Formula Weight 116.07
Percent Purity 97%
Chemical Name or Material 1,4-Dioxane-2,5-dione
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-954-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only