missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane-2,3-diol, 98%

Catalog No. ac438370050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC438370050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC438370050 Supplier Thermo Scientific Chemicals Supplier No. 438370050
Only null left

CAS: 4845-50-5 | C4H8O4 | 120.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4845-50-5
Molecular Formula C4H8O4
Molecular Weight (g/mol) 120.10
MDL Number MFCD00006572
InChI Key YLVACWCCJCZITJ-UHFFFAOYNA-N
Synonym 2,3-dihydroxy-1,4-dioxane, p-dioxane-2,3-diol, 2,3-dihydroxy-1,4-dioxan, dioxanediol, 1,3-diol, 1,4dioxane-2,3-diol, acmc-1al8g, dsstox_cid_18824, dsstox_rid_79404, dsstox_gsid_38824
PubChem CID 96170
IUPAC Name 1,4-dioxane-2,3-diol
SMILES OC1OCCOC1O

Specifications

Melting Point 92°C to 94°C
Color White
CAS Min % 97.5
CAS Max % 100.0
Quantity 5 g
Elemental Analysis 39.10 to 41.10% (C)
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Powder or Crystalline Powder
Assay Percent Range 98%
Formula Weight 120.11
Percent Purity 98%
Chemical Name or Material 1, 4-Dioxane-2, 3-diol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-431-3

RUO – Research Use Only