accessibility menu, dialog, popup

UserName

OHOOHO

CAS RN 4845-50-5

IUPAC Name: 1,4-dioxane-2,3-diol
Synonyms: 1,4-dioxane-2,3-diol
Molecular Weight (g/mol): 120.1
Molecular Formula: C4H8O4
InChi Key: YLVACWCCJCZITJ-UHFFFAOYNA-N
SMILES: OC1OCCOC1O
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)