missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dimethylnaphthalene, MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore 37883100MG
Description
Featured Industries: EnvironmentalSpecifications
| 571-58-4 | |
| 113°C - Closed Cup | |
| C10H6(CH3)2 | |
| -18°C | |
| Analytical Standard | |
| 156.22 |
| 1.016 g/mL (at 25°C) | |
| Neat | |
| MFCD00004037 | |
| 262°C to 264°C (751 mmHg) | |
| 100 mg | |
| Analytical Standard |