missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethoxybenzene 99.0+%, TCI America™

Catalog No. p-7126330
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
D062925G 25 g
D0629500G 500 g
2 options

Catalog No. D062925G

Supplier: TCI America D062925G

Only null left

Chemical Identifiers

CAS 150-78-7
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00008401
InChI Key OHBQPCCCRFSCAX-UHFFFAOYSA-N
Synonym p-dimethoxybenzene, p-methoxyanisole, hydroquinone dimethyl ether, benzene, 1,4-dimethoxy, quinol dimethyl ether, benzene, p-dimethoxy, dimethylhydroquinone ether, dimethyl ether hydroquinone, 1,4-dimethoxybenzol, usaf an-9
PubChem CID 9016
IUPAC Name 1,4-dimethoxybenzene
SMILES COC1=CC=C(OC)C=C1

Specifications

Melting Point 56°C
Color White
Boiling Point 212°C
Quantity 25 g
Formula Weight 138.17
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dimethoxybenzene
EINECSNumber (3)-0582
RTECSNumber CZ6650000
TSCA Yes