missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethoxybenzene, 98%

CAS: 150-78-7 | C8H10O2 | 138.17 g/mol

Supplier:  Thermo Scientific Chemicals A128220E

 View more versions of this product

Catalog No. AAA128220E

Only null left
Explore available promotions

Description

Description

1,4-Dimethoxybenzene is used as Pharmaceutical Intermediate. It is used in some paints and as a diazo dye. It is also used in perfumes and flavors in order to its floral odor. In addition, it is used on greasy skin, and with sulfur to treat acne, or as a dandruff treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dimethoxybenzene is used as Pharmaceutical Intermediate. It is used in some paints and as a diazo dye. It is also used in perfumes and flavors in order to its floral odor. In addition, it is used on greasy skin, and with sulfur to treat acne, or as a dandruff treatment.

Solubility
Soluble in water(0.83g/L at 20°C) and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

1,4-Dimethoxybenzene
54°C to 58°C
212°C
Sweet
MFCD00008401
774605
p-dimethoxybenzene, p-methoxyanisole, hydroquinone dimethyl ether, benzene, 1,4-dimethoxy, quinol dimethyl ether, benzene, p-dimethoxy, dimethylhydroquinone ether, dimethyl ether hydroquinone, 1,4-dimethoxybenzol, usaf an-9
OHBQPCCCRFSCAX-UHFFFAOYSA-N
1,4-dimethoxybenzene
9016
98%
150-78-7
1.053
94°C (201°F)
C8H10O2
2500 g
Light sensitive
Soluble in water(0.83g/L at 20°C) and methanol.
COC1=CC=C(OC)C=C1
138.17
138.17

Chemical Identifiers

150-78-7
138.17
OHBQPCCCRFSCAX-UHFFFAOYSA-N
9016
COC1=CC=C(OC)C=C1
C8H10O2
MFCD00008401
p-dimethoxybenzene, p-methoxyanisole, hydroquinone dimethyl ether, benzene, 1,4-dimethoxy, quinol dimethyl ether, benzene, p-dimethoxy, dimethylhydroquinone ether, dimethyl ether hydroquinone, 1,4-dimethoxybenzol, usaf an-9
1,4-dimethoxybenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315
Causes skin irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 205-771-9

RTECSNumber : CZ6650000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only